Arsenic (III) sulfide ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
221840-25G
221840-5G
BT221840-25GEA
419
USD
BT221840-25G
BT221840-5G
Arsenic (III) sulfide ≥99.999% (trace metals basis)
Arsenic(III) sulphide
CAS: 1303-33-9; EC No: 215-117-4; MDL No: MFCD00003435; RTECS: CG2638000
UN No: UN1557; Haz Class: 6.1; Packing Group: III
12mm & Down Fused Lump ; Molecular Formula: As2S3; MW: 246.03
- RTECS: CG2638000
- 12mm & Down Fused Lump
Formula:
As₂S₃ |
MDL Number:
MFCD00003435 CAS Number: 1303-33-9 EINECS: 215-117-4 UN: 1557 ADR: 6.1,III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...