Sodium hydrogen sulphate monohydrate

Supplier: BeanTown Chemical

Synonyms: Sodium bisulphate monohydrate

221820-1KG 221820-500G 221820-5KG
BT221820-1KGEA 116.95 USD
BT221820-1KG BT221820-500G BT221820-5KG
Sodium hydrogen sulphate monohydrate
Sodium hydrogen sulphate monohydrate

CAS: 10034-88-5; EC No: 231-665-7; MDL No: MFCD00149210; RTECS: VZ1860000 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: NaHSO4·H2O; MW: 138.07 Melting Point: 183° (decomposes) Density (g/mL): 2.103 Moisture Sensitive


  • RTECS: VZ1860000
  • Moisture Sensitive
  • Crystalline

Formula: NaHSO₄·H₂O
MW: 138.08 g/mol
Melting Pt: 183 (dec.)
Density: 2.103 g/mL
MDL Number: MFCD00149210
CAS Number: 10034-88-5
EINECS: 231-665-7
UN: 3260
ADR: 8 III
Merck Index: 13,08658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR