Chlorobenzene ≥99.5% ACS

Supplier: BeanTown Chemical

221810-1L 221810-4L 221810-500ML
BT221810-1LEA 76.5 USD
BT221810-1L BT221810-4L BT221810-500ML
Chlorobenzene ≥99.5% ACS
Chlorobenzene

CAS: 108-90-7; EC No: 203-628-5; MDL No: MFCD00000530; RTECS: CZ0175000 UN No: UN1134; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C6H5Cl; MW: 112.56 Melting Point: -45°; Boiling Point: 132°; Flash point: 27°C (81°F) Density (g/mL): 1.106; Refractive Index: 1.524


  • RTECS: CZ0175000
  • Refractive index: 1.524
  • Liquid

Formula: C₆H₅Cl
Boiling Pt: 132 °C
Melting Pt: –45 °C
Density: 1.106 g/mL
Flash Pt: 27 °C (81 °F)
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR