Ruthenium ≥99.95%, sponge -20 mesh
Supplier: BeanTown Chemical
221770-1G
221770-5G
BT221770-1GEA
150
USD
BT221770-1G
BT221770-5G
Ruthenium ≥99.95%, sponge -20 mesh
Ruthenium
CAS: 7440-18-8; EC No: 231-127-1; MDL No: MFCD00011207
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: Ru; MW: 101.07
Melting Point: 2310°; Boiling Point: 3900°
Density (g/mL): 12.45
- Solid
Formula:
Ru MW: 101.07 Boiling Pt: 3900 °C Melting Pt: 2310 °C Density: 12.45 g/mL |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 EINECS: 231-127-1 ADR: 4.1 III |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...