Cobalt(II) perchlorate hexahydrate
Supplier: BeanTown Chemical
Synonyms:
Cobalt diperchlorate hexahydrate
221670-10G
221670-250G
221670-50G
BT221670-10GEA
27
USD
BT221670-10G
BT221670-250G
BT221670-50G
Cobalt(II) perchlorate hexahydrate
Cobalt(II) perchlorate hexahydrate
CAS: 13478-33-6; EC No: 236-653-5; MDL No: MFCD00150207
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: Co(ClO4)2·6H2O; MW: 365.93
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Cl₂CoO₈·6H₂O MW: 365.93 |
MDL Number:
MFCD00150207 CAS Number: 13478-33-6 EINECS: 236-653-5 UN: 1481 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...