2,3,5-Collidine ≥99%

Supplier: BeanTown Chemical

Synonyms: 2,3,5-Trimethylpyridine

221605-100G 221605-25G 221605-5G
BT221605-100GEA 298 USD
BT221605-100G BT221605-25G BT221605-5G
2,3,5-Collidine ≥99%
2,3,5-Collidine

CAS: 695-98-7; EC No: 211-786-1; MDL No: MFCD00126840 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C8H11N; MW: 121.18 Boiling Point: 184°; Flash point: 73°C (163°F) Density (g/mL): 0.931; Refractive Index: 1.508


  • Refractive index: 1.508
  • Liquid

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 184 °C
Density: 0.931 g/mL
Flash Pt: 73 °C (163 °F)
MDL Number: MFCD00126840
CAS Number: 695-98-7
EINECS: 211-786-1
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR