1,3-Diethylbenzene ≥97%
Supplier: BeanTown Chemical
221545-10ML
221545-2ML
BT221545-10MLEA
239
USD
BT221545-10ML
BT221545-2ML
1,3-Diethylbenzene ≥97%
1,3-Diethylbenzene
CAS: 141-93-5; EC No: 205-511-4; MDL No: MFCD00009260; RTECS: CZ5620000
UN No: UN2049; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C10H14; MW: 134.22
Flash point: 51°C (124°F)
Density (g/mL): 0.864; Refractive Index: 1.496
- RTECS: CZ5620000
- Refractive index: 1.496
- Liquid
Formula:
C₁₀H₁₄ MW: 134.22 g/mol Density: 0.864 g/mL Flash Pt: 51 °C (124 °F) |
MDL Number:
MFCD00009260 CAS Number: 141-93-5 EINECS: 205-511-4 UN: 2049 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...