(2-Methoxyethoxy)methyl chloride ≥94%
Supplier: BeanTown Chemical
Synonyms:
1-Chloromethoxy-2-methoxyethane, 1-Chloro-2,5-dioxahexane, MEM chloride
221540-25G
221540-5G
BT221540-25GEA
150.31
USD
BT221540-25G
BT221540-5G
(2-Methoxyethoxy)methyl chloride ≥94%
(2-Methoxyethoxy)methyl chloride
CAS: 3970-21-6; EC No: 223-589-8; MDL No: MFCD00000888
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: III
Liquid; Linear Formula: CH3OCH2CH2OCH2Cl; MW: 124.57
Boiling Point: 50-52°/13 mmHg; Flash point: 54°C (129°F)
Density (g/mL): 1.091; Refractive Index: 1.427
Moisture Sensitive
- Refractive index: 1.427
- Moisture Sensitive
- Liquid
Formula:
ClCH₂OCH₂CH₂OCH₃ Boiling Pt: 50…52 °C/13 mmHg Density: 1.091 g/mL Flash Pt: 54 °C (129 °F) |
MDL Number:
MFCD00000888 CAS Number: 3970-21-6 EINECS: 223-589-8 UN: 1992 ADR: 3,III |
Specification Test Results
Assay | 94% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...