Barium oxide ≥90%, Technical Grade
Supplier: BeanTown Chemical
221525-1KG
221525-250G
BT221525-1KGEA
151.22
USD
BT221525-1KG
BT221525-250G
Barium oxide ≥90%, Technical Grade
Barium oxide
CAS: 1304-28-5; EC No: 215-127-9; MDL No: MFCD00003453
UN No: UN1884; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: BaO; MW: 153.33
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
BaO MW: 153.33 g/mol |
MDL Number:
MFCD00003453 CAS Number: 1304-28-5 EINECS: 215-127-9 UN: 1884 ADR: 6.1,III |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...