Acetaldehyde ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethanal

221485-100ML 221485-500ML
BT221485-100MLEA 72.17 USD
BT221485-100ML BT221485-500ML
Acetaldehyde ≥98%
Acetaldehyde

CAS: 75-07-0; EC No: 200-836-8; MDL No: MFCD00006991 UN No: UN1089; Haz Class: 3; Packing Group: I Liquid; Molecular Formula: CH3CHO; MW: 44.05 Melting Point: -125°; Boiling Point: 21°; Flash point: -40°C (-40°F) Density (g/mL): 0.785; Refractive Index: 1.332


  • Refractive index: 1.332
  • Liquid

Formula: CH₃CHO
MW: 44.05 g/mol
Boiling Pt: 21 °C
Melting Pt: –125 °C
Density: 0.785 g/mL
Flash Pt: –40 °C (–40 °F)
MDL Number: MFCD00006991
CAS Number: 75-07-0
EINECS: 200-836-8
UN: 1089
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de03379d171eda3c733a039cd16293e8ee4a02e2&vid=573943501bd6753f30bb12e41521b99361988998&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR