Malononitrile ≥99%

Supplier: BeanTown Chemical

221460-100G 221460-2.5KG 221460-500G
BT221460-100GEA 31.93 USD
BT221460-100G BT221460-2.5KG BT221460-500G
Malononitrile ≥99%
Malononitrile

CAS: 109-77-3; EC No: 203-703-2; MDL No: MFCD00001883; RTECS: OO3150000 UN No: UN2647; Haz Class: 6.1; Packing Group: II Solid; Linear Formula: CH2(CHN)2; MW: 66.06 Melting Point: 30-32°; Boiling Point: 220°; Flash point: 86°C (187°F) Density (g/mL): 1.049; Refractive Index: 1.415


  • RTECS: OO3150000
  • Refractive index: 1.415
  • Solid

Formula: C₃H₂N₂
MW: 66.06 g/mol
Boiling Pt: 220 °C
Melting Pt: 30…32 °C
Density: 1.049 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00001883
CAS Number: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR