1,10-Phenanthroline hydrochloride monohydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate

221440-100G 221440-25G 221440-5G
BT221440-100GEA 445 USD
BT221440-100G BT221440-25G BT221440-5G
1,10-Phenanthroline hydrochloride monohydrate ≥99%
1,10-Phenanthroline hydrochloride monohydrate

CAS: 18851-33-7; EC No: 223-325-1; MDL No: MFCD00150061 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C12H8N2·HCl·H2O; MW: 234.68 Melting Point: 224-225°


  • Solid

Formula: C₁₂H₉ClN₂·1H₂O
Melting Pt: 224…225 °C
MDL Number: MFCD00150061
CAS Number: 18851-33-7
EINECS: 223-325-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR