Sodium hexafluorosilicate ≥99%

Supplier: BeanTown Chemical

221430-2KG 221430-500G
BT221430-2KGEA 128.52 USD
BT221430-2KG BT221430-500G
Sodium hexafluorosilicate ≥99%
Sodium hexafluorosilicate

CAS: 16893-85-9; EC No: 240-934-8; MDL No: MFCD00003491; RTECS: VV8410000 UN No: UN2674; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: Na2SiF6; MW: 188.06 Density (g/mL): 2.679; Refractive Index: 1.310


  • RTECS: VV8410000
  • Refractive index: 1.310
  • Crystalline/Powder

Formula: F₆Na₂Si
MW: 188.06
Density: 2.679 g/mL
MDL Number: MFCD00003491
CAS Number: 16893-85-9
EINECS: 240-934-8
UN: 2674
ADR: 6.1 III
Merck Index: 12,08769

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR