2,2-Difluorocyclopropanecarboxylic acid ≥95%
Supplier: BeanTown Chemical
221425-1G
221425-250MG
221425-5G
BT221425-1GEA
393
USD
BT221425-1G
BT221425-250MG
BT221425-5G
2,2-Difluorocyclopropanecarboxylic acid ≥95%
2,2-Difluorocyclopropanecarboxylic acid
CAS: 107873-03-0; MDL No: MFCD04115828
UN No: UN3261; Haz Class: 8; Packing Group: III
Solid; Molecular Formula: C4H4F2O2; MW: 122.07
Melting Point: 60-65°; Boiling Point: 70°/7 mmHg
- Solid
Formula:
C₄H₄F₂O₂ MW: 122.07 g/mol Boiling Pt: 70 °C/7 mmHg Melting Pt: 60…65 °C |
MDL Number:
MFCD04115828 CAS Number: 107873-03-0 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...