Cobalt(II) oxalate dihydrate
Supplier: BeanTown Chemical
Synonyms:
Cobalt(II) oxalate dihydrate
221390-100G
221390-25G
221390-5G
BT221390-100GEA
138
USD
BT221390-100G
BT221390-25G
BT221390-5G
Cobalt(II) oxalate dihydrate
Cobalt (II) oxalate dihydrate
CAS: 5965-38-8; EC No: 212-409-3; MDL No: MFCD00050793
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: CoC2O4·2H2O ; MW: 182.98
Melting Point: 230°
Density (g/mL): 3.021
- Powder
Formula:
C₂CoO₄·2H₂O MW: 182.98 g/mol (146.95 anhy) Melting Pt: 230 °C Density: 3.021 g/mL |
MDL Number:
MFCD03414148 CAS Number: 5965-38-8 EINECS: 212-409-3 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...