Hexabutylditin ≥98%

Supplier: BeanTown Chemical

Synonyms: Hexa-n-butylditin, Hexabutyldistannane, Bis(tributyltin)

221380-10G 221380-250G 221380-50G
BT221380-10GEA 48.49 USD
BT221380-10G BT221380-250G BT221380-50G
Hexabutylditin ≥98%
Hexabutylditin

CAS: 813-19-4; EC No: 212-383-3; MDL No: MFCD00009417 UN No: UN2788; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C24H54Sn2; MW: 580.07 Boiling Point: 197-198°/10 mmHg; Flash point: 124°C (255°F) Density (g/mL): 1.148; Refractive Index: 1.512 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.512
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₂₄H₅₄Sn₂
MW: 580.11 g/mol
Boiling Pt: 197…198 °C/10 mmHg
Density: 1.148 g/mL
Flash Pt: 124 °C (255 °F)
MDL Number: MFCD00009417
CAS Number: 813-19-4
EINECS: 212-383-3
UN: 2788
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR