Ammonium hexachloroplatinate(IV)

Supplier: BeanTown Chemical

Synonyms: Diammonium hexachloroplatinate

221330-1G 221330-5G
BT221330-1GEA 131 USD
BT221330-1G BT221330-5G
Ammonium hexachloroplatinate(IV)
Ammonium hexachloroplatinate(IV)

CAS: 16919-58-7; EC No: 240-973-0; MDL No: MFCD00010886; RTECS: BP5425000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: (NH4)2PtCl6; MW: 443.87 Density (g/mL): 3.07 Hygroscopic


  • RTECS: BP5425000
  • Hygroscopic
  • Powder

Formula: H₈Cl₆N₂Pt
MW: 443.87
Density: 3.07 g/mL
MDL Number: MFCD00010886
CAS Number: 16919-58-7
EINECS: 240-973-0
UN: 3288
ADR: 6.1 III
Merck Index: 12,00582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR