Silver nitrate ≥99.9% ACS
Supplier: BeanTown Chemical
Synonyms:
Lunar caustic, Silver mononitrate, Nitric acid silver salt
221305-100G
221305-25G
221305-5G
BT221305-100GEA
316.44
USD
BT221305-100G
BT221305-25G
BT221305-5G
Silver nitrate ≥99.9% ACS
Silver nitrate
CAS: 7761-88-8; EC No: 231-853-9; MDL No: MFCD00003414; RTECS: VW4725000
UN No: UN1479; Haz Class: 5.1; Packing Group: I
Crystalline; Molecular Formula: AgNO3; MW: 169.87
Melting Point: 212° (decomposes); Boiling Point: 440°
Density (g/mL): 4.35
Light Sensitive
- RTECS: VW4725000
- Light Sensitive
- Crystalline
Formula:
AgNO₃ MW: 169.87 g/mol Boiling Pt: 444 Melting Pt: 210 (dec.) Density: 4.35 g/mL |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 1479 ADR: 5.1 I Merck Index: 12,08661 |
|
Specification Test Results
Assay | 99.9% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...