Benzenesulfonyl chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Benzene sulphochloride
221300-1KG
221300-250G
221300-5KG
BT221300-1KGEA
80
USD
BT221300-1KG
BT221300-250G
BT221300-5KG
Benzenesulfonyl chloride ≥98%
Benzenesulphonyl chloride
CAS: 98-09-9; EC No: 202-636-6; MDL No: MFCD00007426; RTECS: DB8750000
UN No: UN2225; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: C6H5SO2Cl; MW: 176.62
Melting Point: 13-15°; Boiling Point: 251-252°; Flash point: 132°C (270°F)
Density (g/mL): 1.384; Refractive Index: 1.551
Moisture Sensitive
- RTECS: DB8750000
- Refractive index: 1.551
- Moisture Sensitive
- Liquid
Formula:
C₆H₅ClO₂S MW: 176.62 g/mol Boiling Pt: 251…252 °C Melting Pt: 13…15 °C Density: 1.384 g/mL Flash Pt: 132 °C (270 °F) |
MDL Number:
MFCD00007426 CAS Number: 98-09-9 EINECS: 202-636-6 UN: 2225 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...