Bromobenzene ≥99%

Supplier: BeanTown Chemical

Synonyms: Phenylbromide, Phenyl bromide

221285-10KG 221285-2.5KG 221285-500G
BT221285-10KGEA 367 USD
BT221285-10KG BT221285-2.5KG BT221285-500G
Bromobenzene ≥99%
Bromobenzene

CAS: 108-86-1; EC No: 203-623-8; MDL No: MFCD00000055; RTECS: CY9000000 UN No: UN2514; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C6H5Br; MW: 157.01 Melting Point: -31°; Boiling Point: 156°; Flash point: 51°C (124°F) Density (g/mL): 1.491; Refractive Index: 1.559


  • RTECS: CY9000000
  • Refractive index: 1.559
  • Liquid

Formula: C₆H₅Br
MW: 157.01 g/mol
Boiling Pt: 156 °C
Melting Pt: –31 °C
Density: 1.491 g/mL
Flash Pt: 51 °C (124 °F)
MDL Number: MFCD00000055
CAS Number: 108-86-1
EINECS: 203-623-8
UN: 2514
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR