2-Nitrobenzenesulfonyl chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Nitrobenzene-1-sulphonyl chloride
221280-100G
221280-25G
BT221280-100GEA
111
USD
BT221280-100G
BT221280-25G
2-Nitrobenzenesulfonyl chloride ≥97%
2-Nitrobenzenesulphonyl chloride
CAS: 1694-92-4; EC No: 216-907-1; MDL No: MFCD00007430
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: C6H4ClNO4S ; MW: 221.62
Melting Point: 63-67°
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
C₆H₄ClNO₄S MW: 221.62 g/mol Melting Pt: 63…67 °C |
MDL Number:
MFCD00007430 CAS Number: 1694-92-4 EINECS: 216-907-1 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...