Niobium diselenide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Niobium(IV) selenide
221220-10G
221220-50G
BT221220-10GEA
75
USD
BT221220-10G
BT221220-50G
Niobium diselenide ≥99.5%
Niobium diselenide
CAS: 12034-77-4; EC No: 234-811-8; MDL No: MFCD00016228
UN No: UN3283; Haz Class: 6.1; Packing Group: III
5 Micron Powder; Molecular Formula: NbSe2; MW: 250.83
Melting Point:
- 5 Micron Powder
Formula:
NbSe₂ MW: 250.83 Melting Pt: >1316 °C Density: 6.3 g/mL |
MDL Number:
MFCD00016228 CAS Number: 12034-77-4 EINECS: 234-811-8 UN: 3283 ADR: 6.1 III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...