Propionic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
Propanoic acid
221215-1KG
221215-2.5KG
BT221215-1KGEA
42.34
USD
BT221215-1KG
BT221215-2.5KG
Propionic acid ≥99%
Propionic acid
CAS: 79-09-4; EC No: 201-176-3; MDL No: MFCD00002756; RTECS: UE5950000
UN No: UN3463; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: CH3CH2COOH; Molecular Formula: C3H6O2; MW: 74.08
Melting Point: -24 - -23°; Boiling Point: 141-142°; Flash point: 51°C (123°F)
Density (g/mL): 0.992; Refractive Index: 1.386
- RTECS: UE5950000
- Refractive index: 1.386
- Liquid
Formula:
C₃H₆O₂ MW: 74.08 g/mol Boiling Pt: 141…142 °C Melting Pt: –24 – –23 °C Density: 0.992 g/mL Flash Pt: 51 °C (123 °F) |
MDL Number:
MFCD00002756 CAS Number: 79-09-4 EINECS: 201-176-3 UN: 3463 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...