(S)-1-Boc-2-(aminomethyl)pyrrolidine ≥97%
Supplier: BeanTown Chemical
221125-1G
221125-5G
BT221125-1GEA
65
USD
BT221125-1G
BT221125-5G
(S)-1-Boc-2-(aminomethyl)pyrrolidine ≥97%
(S)-1-Boc-2-(aminomethyl)pyrrolidine
CAS: 119020-01-8; MDL No: MFCD03419257
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C10H20N2O2 ; MW: 200.28
- Liquid
Formula:
C₁₀H₂₀N₂O₂ |
MDL Number:
MFCD03419257 CAS Number: 119020-01-8 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...