Methoxyflurane ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,2-Dichloro-1,1-difluoroethyl methyl ether
221090-25G
221090-5G
BT221090-25GEA
307
USD
BT221090-25G
BT221090-5G
Methoxyflurane ≥97%
Methoxyflurane
CAS: 76-38-0; EC No: 200-956-0; MDL No: MFCD00040144; RTECS: KN7820000
UN No: UN3271; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3OCF2CHCl2; Molecular Formula: C3H4Cl2F2O; MW: 164.97
Flash point: 37°C (98°F)
Density (g/mL): 1.440
- RTECS: KN7820000
- Liquid
Formula:
C₃H₄Cl₂F₂O MW: 164.961 g/mol Density: 1.440 g/mL Flash Pt: 37 °C (98 °F) |
MDL Number:
MFCD00040144 CAS Number: 76-38-0 EINECS: 200-956-0 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...