Tris(cyclopentadienyl)gadolinium(III) ≥99.9% (trace rare earth metals basis)

Supplier: BeanTown Chemical

221075-1G 221075-5G
BT221075-1GEA 148 USD
BT221075-1G BT221075-5G
Tris(cyclopentadienyl)gadolinium(III) ≥99.9% (trace rare earth metals basis)
Tris(cyclopentadienyl)gadolinium(III)

CAS: 1272-21-5; EC No: 215-046-9; MDL No: MFCD00145476 UN No: UN3181; Haz Class: 4.1; Packing Group: III Powder; Linear Formula: Gd(C5H5)3; Molecular Formula: C15H15Gd ; MW: 352.54 Melting Point: 295° (decomposes); Flash point: 35°C (95°F) Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: C₁₅H₁₅Gd
MW: 352.535 g/mol
Melting Pt: 295 °C (dec.)
Flash Pt: 35 °C (95 °F)
MDL Number: MFCD00145476
CAS Number: 1272-21-5
EINECS: 215-046-9
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR