(Pentamethylcyclopentadienyl)zirconium trichloride ≥99%
Supplier: BeanTown Chemical
CAS: 75181-07-6; MDL No: MFCD00070444
UN No: UN3096; Haz Class: 8 (4.3); Packing Group: II
Crystalline, packaged in ampules; Molecular Formula: C10H15Cl3Zr ; MW: 332.81
Melting Point: 230° (decomposes)
Moisture Sensitive
- Moisture Sensitive
- Crystalline, packaged in ampules
Formula:
C₁₀H₁₅Cl₃Zr Melting Pt: 230 °C (dec.) |
MDL Number:
MFCD00070444 CAS Number: 75181-07-6 UN: 3096 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...