3-Chloro-1-propanol ≥98%

Supplier: BeanTown Chemical

221015-100G 221015-25G 221015-500G
BT221015-100GEA 68 USD
BT221015-100G BT221015-25G BT221015-500G
3-Chloro-1-propanol ≥98%
3-Chloro-1-propanol

CAS: 627-30-5; EC No: 210-992-9; MDL No: MFCD00002943; RTECS: UA8930000 UN No: UN2849; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: Cl(CH2)3OH; Molecular Formula: C3H7ClO; MW: 94.54 Boiling Point: 160-162°; Flash point: 75°C (167°F) Density (g/mL): 1.131; Refractive Index: 1.445


  • RTECS: UA8930000
  • Refractive index: 1.445
  • Liquid

Formula: C₃H₇ClO
Boiling Pt: 160…162 °C
Density: 1.131 g/mL
Flash Pt: 75 °C (167 °F)
MDL Number: MFCD00002943
CAS Number: 627-30-5
EINECS: 210-992-9
UN: 2849
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR