Pentafluorobenzyl bromide ≥98%
Supplier: BeanTown Chemical
Synonyms:
α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide
221005-100G
221005-25G
221005-5G
BT221005-100GEA
806.27
USD
BT221005-100G
BT221005-25G
BT221005-5G
Pentafluorobenzyl bromide ≥98%
Pentafluorobenzyl bromide
CAS: 1765-40-8; EC No: 217-182-4; MDL No: MFCD00000299
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C7H2BrF5 ; MW: 260.99
Melting Point: 19-20°; Boiling Point: 174-175°; Flash point: 83°C (181°F)
Density (g/mL): 1.728; Refractive Index: 1.471
- Refractive index: 1.471
- Liquid
Formula:
(C₆F₅)CH₂Br MW: 260.99 g/mol Boiling Pt: 174…175 °C Melting Pt: 19…20 °C Density: 1.728 g/mL Flash Pt: 83 °C (181 °F) |
MDL Number:
MFCD00000299 CAS Number: 1765-40-8 EINECS: 217-182-4 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...