(±)-3-Quinuclidinol ≥98%

Supplier: BeanTown Chemical

Synonyms: (±)-Quinuclidin-3-ol, (±)-3-Hydroxyquinuclidine

220990-100G 220990-25G 220990-5G
BT220990-100GEA 370 USD
BT220990-100G BT220990-25G BT220990-5G
(±)-3-Quinuclidinol ≥98%
(±)-3-Quinuclidinol

CAS: 1619-34-7; EC No: 216-578-4; MDL No: MFCD00151326; RTECS: VD6191700 UN No: UN3263; Haz Class: 8; Packing Group: III Powder; Molecular Formula: C7H13NO; MW: 127.18 Melting Point: 220-223° Air Sensitive


  • RTECS: VD6191700
  • Air Sensitive
  • Powder

Formula: C₇H₁₃NO
MW: 127.19 g/mol
Melting Pt: 220…223 °C
MDL Number: MFCD00151326
CAS Number: 1619-34-7
EINECS: 216-578-4
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR