Barium hydroxide octahydrate ≥97%

Supplier: BeanTown Chemical

220955-10KG 220955-2.5KG 220955-500G
BT220955-10KGEA 456 USD
BT220955-10KG BT220955-2.5KG BT220955-500G
Barium hydroxide octahydrate ≥97%
Barium hydroxide octahydrate

CAS: 12230-71-6; EC No: 241-234-5; MDL No: MFCD00149152; RTECS: CQ9200000 UN No: UN3262; Haz Class: 8; Packing Group: III Solid; Linear Formula: Ba(OH)2·8H2O; MW: 315.46 Melting Point: 78° Air Sensitive , Hygroscopic


  • RTECS: CQ9200000
  • Air Sensitive, Hygroscopic
  • Solid

Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR