Magnesium trifluoromethanesulphonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Magnesium trifluoromethanesulphonate, Magnesium triflate

220925-100G 220925-25G 220925-5G
BT220925-100GEA 323 USD
BT220925-100G BT220925-25G BT220925-5G
Magnesium trifluoromethanesulphonate ≥98%
Magnesium trifluoromethanesulphonate

CAS: 60871-83-2; MDL No: MFCD00042601 UN No: UN1759; Haz Class: 8; Packing Group: III Powder; Linear Formula: (CF3SO3)2Mg; MW: 322.43 Melting Point: 300° Hygroscopic


  • Hygroscopic
  • Powder

Formula: C₂F₆MgO₆S₂
MW: 322.45
Melting Pt: >300 °C
MDL Number: MFCD00042601
CAS Number: 60871-83-2
ADR: 8 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR