1-Butanol ≥99.4% ACS
Supplier: BeanTown Chemical
Synonyms:
1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
220845-1L
220845-4L
220845-500ML
BT220845-1LEA
47.64
USD
BT220845-1L
BT220845-4L
BT220845-500ML
1-Butanol ≥99.4% ACS
1-Butanol
CAS: 71-36-3; EC No: 200-751-6; MDL No: MFCD00002964; RTECS: EO1400000
UN No: UN1120; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3(CH2)3OH; Molecular Formula: C4H10O; MW: 74.12
Melting Point: -90°; Boiling Point: 116-118°; Flash point: 35°C (95°F)
Density (g/mL): 0.81; Refractive Index: 1.399
Hygroscopic
- RTECS: EO1400000
- Refractive index: 1.399
- Hygroscopic
- Liquid
Formula:
H₃C(CH₂)₂CH₂OH MW: 74.12 g/mol Boiling Pt: 116…118 °C Melting Pt: –90 °C Density: 0.81 g/mL Flash Pt: 35 °C (95 °F) |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III |
Specification Test Results
Assay | 99.4% min. |
Color (APHA) | ≤10 |
Residue after evaporation | ≤0.005% |
Titrable acid | ≤0.0008meq/g |
Butyl ether | ≤0.2% |
Water | ≤0.1% |
Isobutanol | ≤0.10% |
Carbonyl compounds (as butyraldehyde) | ≤0.01% |
Specific gravity at 25°C | 0.807-0.809 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...