Nickel(II) carbonate basic hydrate ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Nickel(II) carbonate hydroxide hydrate, Basic nickel carbonate
220815-1KG
220815-250G
BT220815-1KGEA
131
USD
BT220815-1KG
BT220815-250G
Nickel(II) carbonate basic hydrate ≥99.5% (trace metals basis)
Nickel(II) carbonate basic hydrate
CAS: 12607-70-4; EC No: 240-408-8; MDL No: MFCD00016262; RTECS: QR6260000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: NiCO3·2Ni(OH)2·xH2O; MW: 304.11 (anhydrous)
- RTECS: QR6260000
- Powder
Formula:
CH₄Ni₃O₇·H₂O |
MDL Number:
MFCD00150262 CAS Number: 39430-27-8 EINECS: 240-408-8 ADR: 9,III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...