Sodium chlorate ≥99%

Supplier: BeanTown Chemical

220795-1KG 220795-250G
BT220795-1KGEA 41.55 USD
BT220795-1KG BT220795-250G
Sodium chlorate ≥99%
Sodium chlorate

CAS: 7775-09-9; EC No: 231-887-4; MDL No: MFCD00003479 UN No: UN1495; Haz Class: 5.1; Packing Group: II Crystalline; Linear Formula: NaClO3; MW: 106.45 Melting Point: 248-261° Density (g/mL): 2.49


  • Crystalline

Formula: ClNaO₃
Melting Pt: 248…261 °C
Density: 2.49 g/mL
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR