Tellurium tetrachloride ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Tellurium(IV) chloride
220775-100G
220775-25G
BT220775-100GEA
391
USD
BT220775-100G
BT220775-25G
Tellurium tetrachloride ≥99.9%
Tellurium tetrachloride
CAS: 10026-07-0; EC No: 233-055-6; MDL No: MFCD00011262; RTECS: WY2635000
UN No: UN3260; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: TeCl4; MW: 269.41
Melting Point: 224°; Boiling Point: 380°
Density (g/mL): 3.26
Moisture Sensitive
- RTECS: WY2635000
- Moisture Sensitive
- Crystalline
Formula:
Cl₄Te MW: 269.41 g/mol Boiling Pt: 380 Melting Pt: 224 Density: 3.26 g/mL |
MDL Number:
MFCD00011262 CAS Number: 10026-07-0 EINECS: 233-055-6 UN: 3260 ADR: 8 II Merck Index: 12,09279 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...