Triethylsilane ≥98%

Supplier: BeanTown Chemical

220745-10G 220745-250G 220745-50G
BT220745-10GEA 41.14 USD
BT220745-10G BT220745-250G BT220745-50G
Triethylsilane ≥98%
Triethylsilane

CAS: 617-86-7; EC No: 210-535-3; MDL No: MFCD00009018 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3CH2)3SiH; Molecular Formula: C6H16Si; MW: 116.28 Melting Point: -157°; Boiling Point: 107-108°; Flash point: -6°C (21°F) Density (g/mL): 0.728; Refractive Index: 1.412 Moisture Sensitive


  • Refractive index: 1.412
  • Moisture Sensitive
  • Liquid

Formula: C₆H₁₆Si
MW: 116.28 g/mol
Boiling Pt: 107
Melting Pt: -156.9
Density: 0.728 g/mL
Flash Pt: –6 °C (21 °F)
MDL Number: MFCD00009018
CAS Number: 617-86-7
EINECS: 210-535-3
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR