Fluorobenzene ≥99%

Supplier: BeanTown Chemical

220735-1KG 220735-250G
BT220735-1KGEA 205 USD
BT220735-1KG BT220735-250G
Fluorobenzene ≥99%
Fluorobenzene

CAS: 462-06-6; EC No: 207-321-7; MDL No: MFCD00000280; RTECS: DA0800000 UN No: UN2387; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C6H5F; MW: 96.10 Melting Point: -42°; Boiling Point: 85°; Flash point: -15°C (5°F) Density (g/mL): 1.157; Refractive Index: 1.465


  • RTECS: DA0800000
  • Refractive index: 1.465
  • Liquid

Formula: C₆H₅F
Boiling Pt: 85 °C
Melting Pt: –42 °C
Density: 1.157 g/mL
Flash Pt: –15 °C (5 °F)
MDL Number: MFCD00000280
CAS Number: 462-06-6
EINECS: 207-321-7
UN: 2387
ADR: 3,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR