1-Butylimidazole ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-Butyl-1H-imidazole, 1-Butyl-1H-imidazole, N-Butylimidazole
220705-100G
220705-25G
BT220705-100GEA
86
USD
BT220705-100G
BT220705-25G
1-Butylimidazole ≥99%
1-Butylimidazole
CAS: 4316-42-1; EC No: 224-335-9; MDL No: MFCD00042753
UN No: UN2690; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C7H12N2; MW: 124.18
Boiling Point: 114-116°; Flash point:
- Refractive index: 1.480
- Liquid
Formula:
C₇H₁₂N₂ MW: 124.19 g/mol Boiling Pt: 114…116 °C Density: 0.945 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00042753 CAS Number: 4316-42-1 EINECS: 224-335-9 UN: 2690 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...