Cadmium oxide ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Cadmium (II) oxide
220675-100G
220675-25G
220675-500G
BT220675-100GEA
240
USD
BT220675-100G
BT220675-25G
BT220675-500G
Cadmium oxide ≥99.995% (trace metals basis)
Cadmium oxide
CAS: 1306-19-0; EC No: 215-146-2; MDL No: MFCD00010921
UN No: UN2570; Haz Class: 6.1; Packing Group: I
Powder; Molecular Formula: CdO; MW: 128.41
Density (g/mL): 8.15
- Powder
Formula:
CdO MW: 128.41 g/mol Density: 8.15 g/mL |
MDL Number:
MFCD00010921 CAS Number: 1306-19-0 EINECS: 215-146-2 UN: 2570 ADR: 6.1,I |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...