Benzene-[D₆] ≥99.5% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: Benzene-[²H₆]

220640-10G 220640-50G
BT220640-10GEA 95.78 USD
BT220640-10G BT220640-50G
Benzene-[D₆] ≥99.5% (Isotopic)
Benzene-[D₆]

CAS: 1076-43-3; EC No: 214-061-8; MDL No: MFCD00003010 UN No: UN1114; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C6D6; MW: 84.15 Melting Point: 6.8°; Boiling Point: 79.1°; Flash point: -11°C (12°F) Density (g/mL): 0.95; Refractive Index: 1.497 Moisture Sensitive


  • Refractive index: 1.497
  • Moisture Sensitive
  • Liquid

Formula: C₆D₆
Boiling Pt: 79.1 °C
Melting Pt: 6.8 °C
Density: 0.95 g/mL
Flash Pt: –11 °C (12 °F)
MDL Number: MFCD00003010
CAS Number: 1076-43-3
EINECS: 214-061-8
UN: 1114
ADR: 3,II

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR