2-Iodofluorene ≥98%
Supplier: BeanTown Chemical
220620-1G
220620-5G
BT220620-1GEA
34
USD
BT220620-1G
BT220620-5G
2-Iodofluorene ≥98%
2-Iodofluorene
CAS: 2523-42-4; MDL No: MFCD00019049
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: C13H9I; MW: 292.12
Melting Point: 126-129°
- Solid
Formula:
C₁₃H₉I Melting Pt: 126…129 °C |
MDL Number:
MFCD00019049 CAS Number: 2523-42-4 ADR: 9,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...