Diisopropyl azodicarboxylate ≥94%

Supplier: BeanTown Chemical

Synonyms: Diisopropyl diazene-1,2-dicarboxylate

220615-100G 220615-25G
BT220615-100GEA 126.4 USD
BT220615-100G BT220615-25G
Diisopropyl azodicarboxylate ≥94%
Diisopropyl azodicarboxylate

CAS: 2446-83-5; EC No: 219-502-8; MDL No: MFCD00008875 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Linear Formula: (CH3)2CHO2CN=N-CO2CH(CH3)2; Molecular Formula: C8H14N2O4; MW: 202.21 Boiling Point: 75°/0.25mm Hg; Flash point: 103°C (223°F) Density (g/mL): 1.027; Refractive Index: 1.420 Light Sensitive


  • Refractive index: 1.420
  • Light Sensitive
  • Liquid

Formula: C₈H₁₄N₂O₄
MW: 202.21 g/mol
Boiling Pt: 75 °C/0.25mm Hg
Density: 1.027 g/mL
Flash Pt: 103 °C (223 °F)
MDL Number: MFCD00008875
CAS Number: 2446-83-5
EINECS: 219-502-8
ADR: 9,III

Order Now

Specification Test Results

Assay 94%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR