Hydroxylammonium sulfate ≥99%

Supplier: BeanTown Chemical

220595-100G 220595-25G 220595-500G
BT220595-100GEA 30 USD
BT220595-100G BT220595-25G BT220595-500G
Hydroxylammonium sulfate ≥99%
Hydroxylammonium sulphate

CAS: 10039-54-0; EC No: 233-118-8; MDL No: MFCD00044869; RTECS: NC5425000 UN No: UN2865; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: NH2OH·0.5H2SO4; MW: 164.14 Melting Point: 170° (decomposes) Density (g/mL): 1.88


  • RTECS: NC5425000
  • Crystalline

Formula: H₈N₂O₆S
Melting Pt: 170 °C (dec.)
Density: 1.88 g/mL
MDL Number: MFCD00044869
CAS Number: 10039-54-0
EINECS: 233-118-8
UN: 2865
ADR: 8,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR