Aluminium fluoride, anhydrous ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Aluminum fluoride

220580-100G 220580-25G 220580-5G
BT220580-100GEA 1388 USD
BT220580-100G BT220580-25G BT220580-5G
Aluminium fluoride, anhydrous ≥99.5%
Aluminium fluoride

CAS: 7784-18-1; EC No: 232-051-1; MDL No: MFCD00003426 UN No: UN3260; Haz Class: 8; Packing Group: II 3-12mm Sintered Lump; Molecular Formula: AlF3 ; MW: 83.98 Melting Point: 1291° Density (g/mL): 3.1 Hygroscopic


  • Hygroscopic
  • 3-12mm Sintered Lump

Formula: AlF₃
MW: 83.98 g/mol
Melting Pt: 1040
Density: 3.1 g/mL
MDL Number: MFCD00003426
CAS Number: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8 II
Merck Index: 12,00351

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR