Nickel acetate tetrahydrate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate
220520-25G
220520-5G
BT220520-25GEA
418
USD
BT220520-25G
BT220520-5G
Nickel acetate tetrahydrate ≥99.999% (trace metals basis)
Nickel acetate tetrahydrate
CAS: 6018-89-9; EC No: 206-761-7; MDL No: MFCD00066973; RTECS: QR6126000
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline; Linear Formula: Ni(OCOCH3)2·4H2O; MW: 248.84
Density (g/mL): 1.798
- RTECS: QR6126000
- Crystalline
Formula:
C₄H₆NiO₄·4H₂O MW: 248.84 g/mol Boiling Pt: 16 Density: 1.798 g/mL |
MDL Number:
MFCD00066973 CAS Number: 6018-89-9 EINECS: 206-761-7 ADR: 9 III Merck Index: 12,06583 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...