Tungsten dichloride dioxide ≥99%
Supplier: BeanTown Chemical
220490-10G
220490-50G
BT220490-10GEA
201
USD
BT220490-10G
BT220490-50G
Tungsten dichloride dioxide ≥99%
Tungsten dichloride dioxide
CAS: 13520-76-8; EC No: 236-862-1; MDL No: MFCD00054136
UN No: UN3260; Haz Class: 8; Packing Group: II
Powder; Linear Formula: WO2Cl2; MW: 286.75
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
Cl₂O₂W MW: 286.75 g/mol |
MDL Number:
MFCD00054136 CAS Number: 13520-76-8 EINECS: 236-862-1 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...