Gold trichloride ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Gold(III) chloride, Gold (III) chloride

220415-1G 220415-5G
BT220415-1GEA 172.22 USD
BT220415-1G BT220415-5G
Gold trichloride ≥99.99% (trace metals basis)
Gold trichloride

CAS: 13453-07-1; EC No: 236-623-1; MDL No: MFCD00014172; RTECS: MD5420000 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Molecular Formula: AuCl3; MW: 303.33 Density (g/mL): 3.9 Hygroscopic


  • RTECS: MD5420000
  • Hygroscopic
  • Crystalline

Formula: AuCl₃
MW: 303.32 g/mol
Density: 3.9 g/mL
MDL Number: MFCD00014172
CAS Number: 13453-07-1
EINECS: 236-623-1
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR