Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97%

Supplier: BeanTown Chemical

220370-1G 220370-25G 220370-5G
BT220370-1GEA 21 USD
BT220370-1G BT220370-25G BT220370-5G
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97%
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride

CAS: 68641-49-6; MDL No: MFCD00010077; RTECS: SZ5871000 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: C6H8ClN2O5P; MW: 254.56 Melting Point: 191° (decomposes) Moisture Sensitive


  • RTECS: SZ5871000
  • Moisture Sensitive
  • Crystalline

Formula: C₆H₈ClN₂O₅P
Melting Pt: 191 °C (dec.)
MDL Number: MFCD00010077
CAS Number: 68641-49-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR