Chlorodiphenylphosphine ≥98%
Supplier: BeanTown Chemical
Synonyms:
DPPC, Diphenylchlorophosphine
220355-10G
220355-250G
220355-50G
BT220355-10GEA
30
USD
BT220355-10G
BT220355-250G
BT220355-50G
Chlorodiphenylphosphine ≥98%
Chlorodiphenylphosphine
CAS: 1079-66-9; EC No: 214-093-2; MDL No: MFCD00000529
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: (C6H5)2PCl; Molecular Formula: C12H10ClP; MW: 220.64
Boiling Point: 320°; Flash point: 112°C (234°F)
Density (g/mL): 1.229; Refractive Index: 1.636
Air Sensitive, Moisture Sensitive
- Refractive index: 1.636
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₁₂H₁₀ClP MW: 220.64 g/mol Boiling Pt: 320 °C Density: 1.229 g/mL Flash Pt: 112 °C (234 °F) |
MDL Number:
MFCD00000529 CAS Number: 1079-66-9 EINECS: 214-093-2 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...